Learn More
Goat anti-Mouse IgM Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen 31440
259.35 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : PN
Additional Details : Weight : 1.81440kg
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31440 has been successfully used in Western blot, and ICC applications. Antibody Specificity: This antibody reacts with the heavy chain portion of mouse IgM, based on immunoelectrophoresis. No antibody was detected against IgG or non-immunoglobulin serum proteins. However, this antibody may cross-react with IgM from other species Restoration and Storage: Store product at 4°C until opened. Restore with 2 mL distilled water (0.8 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Mouse IgM | |
Polyclonal | |
HRP | |
Goat | |
2 mL | |
Secondary | |
4° C | |
IgM |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.8 mg/mL | |
PBS with 15mg/mL BSA and no preservative; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Mouse | |
Lyophilized |