Learn More
Goat anti-Mouse IgM Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184383
Code : LD
Additional Details : Weight : 1.24740kg
Description
This antibody has been preabsorbed and shows minimal cross-reactivity with other mouse immunoglobulin classes. Goat anti Mouse IgM antibody recognizes murine IgM and is cross-adsorbed to reduce reactivity with other mouse heavy or light chains.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Mouse IgM | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Mouse | |
Liquid |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.01% thimerosal | |
Purified mouse IgM | |
1 mg | |
Secondary | |
4° C, do not freeze | |
IgG |