missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 633, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A21046
Code : 50
Additional Details : Weight : 0.00450kg
Description
Product will be shipped at Room Temperature.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Mouse IgM (Heavy chain) Cross-Adsorbed | |
Polyclonal | |
Alexa Fluor 633 | |
Goat | |
purified | |
RUO | |
Mouse | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Mouse Mu immunoglobulin | |
500 μg | |
Secondary | |
4° C, store in dark | |
IgG |