missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Mouse IgM (H+L) Secondary Antibody, DyLight 650, Novus Biologicals™

Product Code. 18405685 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405685 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405685 Supplier Novus Biologicals Supplier No. NBP175203C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

IgM (H+L) Polyclonal antibody specifically detects IgM (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM (H+L)
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot : 1:100-1:2000, ELISA : 1:100-1:2000, Immunohistochemistry : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500, Immunodiffusion, Immunoassay
Formulation 50mM Sodium Borate
Host Species Goat
Immunogen Purified Mouse IgM, whole molecule
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.