missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG3 Secondary Antibody, PE, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68520PE
This item is not returnable.
View return policy
Description
IgG3 Polyclonal antibody specifically detects IgG3 in Mouse samples. It is validated for ELISA
Specifications
| IgG3 | |
| Polyclonal | |
| PBS | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Mouse | |
| Purified |
| ELISA | |
| PE | |
| DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809 | |
| Mouse IgG3 paraproteins | |
| 0.5 mL | |
| Immunology | |
| 3502 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction