missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG3 Heavy Chain Secondary Antibody, DyLight 488, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7523G
This item is not returnable.
View return policy
Description
IgG3 Heavy Chain Polyclonal antibody specifically detects IgG3 Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG3 Heavy Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Mouse IgG3 | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction