missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG2c Heavy Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB7563
This item is not returnable.
View return policy
Description
IgG2c Heavy Chain Polyclonal antibody specifically detects IgG2c Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| IgG2c Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Affinity Purified | |
| RUO | |
| By immunoelectrophoresis and ELISA this reacts specifically with mouse IgG2c. This may cross react with IgG2c from other species. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:10000, ELISA, Immunohistochemistry 1:200- 1:2000, Immunocytochemistry/Immunofluorescence 1:200- 1:2000, Immunohistochemistry-Paraffin | |
| Mouse IgG2c | |
| 1 mL | |
| Secondary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction