missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Mouse IgG2a Cross-Adsorbed Secondary Antibody, PE

Product Code. 10056673
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10056673 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10056673 Supplier Invitrogen™ Supplier No. 31863

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31863 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31863 reacts with the heavy chains of mouse IgG2a, but not with heavy chains of other mouse IgG subclasses or with light chains of most mouse immunoglobulins. This antibody does not react against mouse IgM, or against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human, bovine, and rabbit serum proteins. However, these antibodies may cross-react with immunoglobulins from other species. Phycobiliprotein Concentration: 0.5 mg/mL (determined by absorption = 82.0 at 566 nm for a 1% solution for only those R-PE molecules to which at least one molecule of active antibody is bound) Phycoerythrin Source: Seaweed Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. RPE Amax= 490 nm, 545 nm, and 566 nm; Emax= 580 nm. Reconstitute with 0.5 mL of distilled water (0.5 mg/mL after restoration). Country of Origin: USA.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG2a Cross-Adsorbed
Applications Flow Cytometry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate PE
Formulation PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Host Species Goat
Purification Method Antigen affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C
Product Type Secondary Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.