missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Mouse IgG1 Superclonal™ Secondary Antibody

Goat Recombinant Superclonal Secondary Antibody

Brand:  Invitrogen™ SA5-10373

Product Code. 30485026

  • £193.00 / 250µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Goat anti-Mouse IgG1 Recombinant Superclonal™ Secondary Antibody specifically detects the Fc region of mouse IgG1 with no cross-reactivity to mouse IgG2a, mouse IgG2b or immunoglobulins from other species. The sensitivity and specificity of each lot is confirmed using ELISA. Superclonal™ secondary antibodies are produced using a multifaceted clonal selection process that involves several phenotypic screens to ensure enhanced specificity, animal origin free formulation and lot-to-lot consistency. It is recommended that the antibody is carefully titrated for optimal performance in the assay of interest.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IgG1
Recombinant Superclonal
Unconjugated
Goat
Affinity chromatography
RUO
Mouse
Secondary Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.0 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Purified Mouse IgG.
250 μL
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.