Learn More
Invitrogen™ Goat anti-Mouse IgG1 Superclonal™ Secondary Antibody, Alexa Fluor™ Plus 647
Goat Recombinant Superclonal Secondary Antibody
Brand: Invitrogen™ SA5-10373-AFP647
Description
Goat anti-Mouse IgG1, Superclonal™ Recombinant Secondary Antibody, Alexa Fluor™ Plus 647 specifically detects the Fc region of mouse IgG1 with no cross reactivity to Mouse IgG2a or Mouse IgG2b. Minimal cross reactivity was observed with Mouse IgG3, Human IgG, Rat IgG and Rat IgA. The sensitivity and specificity of each lot of Goat anti-Mouse IgG1, Superclonal™ Recombinant Secondary Antibody is confirmed using ELISA and then taken up for conjugation. Superclonal™ recombinant secondary antibodies are produced using a multifaceted clonal selection process that involves several phenotypic screens to ensure enhanced specificity, animal origin free formulation and lot-to-lot consistency. Alexa Fluor™ Plus conjugated recombinant antibodies provide bright signals and high signal-to-noise for your critical experiments. It is recommended that the antibody is carefully titrated for optimal performance in the assay of interest.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Mouse IgG1 | |
| Recombinant Superclonal | |
| Alexa Fluor Plus 647 | |
| Goat | |
| Affinity chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1.0 mg/mL | |
| Proprietary buffer with 0.008 Bromonitrodioxane, 0.008 Methylisothiazolone; pH 6.3-7.3 | |
| Purified Mouse IgG. | |
| 250 μL | |
| Secondary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.