Learn More
Invitrogen™ Goat anti-Mouse IgG1 Secondary Antibody, PE-Texas Red
Goat Polyclonal Secondary Antibody
Brand:  Invitrogen™ M32017
Product Code. 10291853
  • £247.00 / 500µL
For Research Use Only. All usage must comply with product instructions.
Description
Description
Goat anti-Mouse IgG1 Secondary Antibody for Flow
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse IgG1
Polyclonal
PBS with 0.1% sodium azide
IgG gamma 1.
500 μL
Secondary
4°C, store in dark
Liquid
Flow Cytometry
PE-Texas Red
Goat
Purified
RUO
Mouse
Secondary Antibody