Learn More
Invitrogen™ Goat anti-Mouse IgG1 Secondary Antibody, HRP
Goat Polyclonal Secondary Antibody
Brand:  Invitrogen™ PA174421
Product Code. 11554240
  • £363.00 / 1mL
Description
This antibody does not react with human. Goat anti Mouse IgG1 antibody recognizes Mouse IgG1.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse IgG1
Polyclonal
PBS with 50% glycerol and 0.0095% Methylisothiazolone; pH 7.4
Mouse IgG1 paraproteins.
1 mL
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
HRP
Goat
Affinity chromatography
RUO
Mouse
Secondary Antibody