missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Mouse IgG1 Heavy Chain Secondary Antibody, DyLight 650, Novus Biologicals™

Product Code. 18420515 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420515 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420515 Supplier Novus Biologicals Supplier No. NB7508C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

IgG1 Heavy Chain Polyclonal antibody specifically detects IgG1 Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1 Heavy Chain
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot : 1:1000 - 1:30000, ELISA : 1:1000- 1:30000, Immunohistochemistry : 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence : 1:200 - 1:2000, Immunohistochemistry-Paraffin : 1:200 - 1:2000
Formulation 50 mM Sodium Borate
Host Species Goat
Immunogen Mouse IgG1
Purification Method Immunogen affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.