missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG1 Heavy Chain Secondary Antibody, DyLight 488, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG1 Heavy Chain Polyclonal antibody specifically detects IgG1 Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | IgG1 Heavy Chain |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Dilution | Western Blot : 1:1000 - 1:30000, ELISA : 1:1000- 1:30000, Immunohistochemistry : 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence : 1:200 - 1:2000, Immunohistochemistry-Paraffin : 1:200 - 1:2000 |
| Formulation | 50 mM Sodium Borate |
| Host Species | Goat |
| Immunogen | Mouse IgG1 |
| Purification Method | Immunogen affinity purified |
| Quantity | 1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?