missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG/IgA/IgM/IgD (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69510
This item is not returnable.
View return policy
Description
IgG/IgA/IgM/IgD (H+L) Polyclonal antibody specifically detects IgG/IgA/IgM/IgD (H+L) in Mouse samples. It is validated for Immunoprecipitation, Immunoelectrophoresis
Specifications
| IgG/IgA/IgM/IgD (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Whole Antiserum | |
| RUO | |
| Reconstitute by adding 1 mL sterile distilled water. Working dilutions are prepared by adding sterile phosphate buffered saline (PBS, pH 7.2). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunoprecipitation, Immunoelectrophoresis | |
| LYOPH | |
| Whole antiserum | |
| Purified polyclonal mouse IgG and pools of homogenous IgG, IgA, IgM and IgD. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Secondary | |
| Mouse | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction