Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, Alexa Fluor™ 488
Goat Recombinant Superclonal Secondary Antibody
£70.50 - £328.00
Specifications
| Antigen | Mouse IgG (H+L) |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Recombinant Superclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15328478
|
Invitrogen™
A28175 |
1 mg |
£328.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15607878
|
Invitrogen™
A28175SAMPLE |
50 μg |
£70.50
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with rabbit, rat, human, bovine, guinea pig and donkey IgG is observed. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
| Mouse IgG (H+L) | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Recombinant Superclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Recombinant full-length protein. | |
| Secondary Antibody |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| Alexa Fluor 488 | |
| Goat | |
| RUO | |
| PBS with 5mM sodium azide | |
| Secondary | |
| Gravity column chromatography |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.