missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
£86.00
Specifications
| Antigen | IgG (H+L) |
|---|---|
| Concentration | 4.5 mg/mL |
| Applications | Immunoassay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay.Specifications
| IgG (H+L) | |
| Immunoassay | |
| Unconjugated | |
| RUO | |
| Mouse | |
| IgG | |
| Affinity purified | |
| Based on IEP, this antibody reacts with heavy gamma chains on mouse IgG and light chains on all mouse immunoglobulins. This antibody has been pre-adsorbed against bovine, goat, human, rabbit, and rat IgG. |
| 4.5 mg/mL | |
| Polyclonal | |
| Goat | |
| Adaptive Immunity, Immunology | |
| Purified Mouse IgG (H&L) | |
| Secondary | |
| Store at 4°C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20°C. Avoid repeated freeze/thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts