missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Mouse IgG (H+L) Secondary Antibody, PE, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NB7535PE

Product Code. 30230853

  • £393.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG (H+L)
Polyclonal
PBS
This Goat anti-Mouse IgG (H+L) Secondary Antibody was developed against mouse IgG-heavy and light chain.
1 mL
Adaptive Immunity, Immunology
Mouse
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
PE
Goat
Immunogen affinity purified
RUO
Secondary
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.