Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Secondary Antibody, DyLight™ 800 4X PEG
Description
Product SA5-35521 reacts with the heavy chains of Mouse IgG and with the light chains common to most Mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol without and store at -20°C. Note: not compatible with sodium azide. Mole Dye/Mole Protein Ratio is lot-dependent. DyLight 800-4xPEG has an excitation (nm) of 783 nm (Amax) and an emission (nm) of 797 nm (Emax) in PBS. The emission spectra for DyLight™ 800-4xPEG conjugate match the principal output wavelengths of most common fluorescence instrumentation.
Specifications
Specifications
| Antigen | Mouse IgG (H+L) |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | DyLight 800 4X PEG |
| Formulation | Proprietary buffer with no preservative |
| Host Species | Goat |
| Immunogen | Purified Mouse IgG, whole molecule. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.