Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Secondary Antibody, Biotin
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ 31800
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31800 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31800 reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 2.0 mL of distilled water. Country of Origin: USA.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Mouse IgG (H+L) | |
| Polyclonal | |
| Biotin | |
| Goat | |
| 2 mL | |
| Secondary | |
| 4°C | |
| Lyophilized | |
| Responsibly packaged |
| ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.3 mg/mL | |
| PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
| Antigen affinity chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
Safety and Handling
- Goat anti-Mouse IgG (H+L) Secondary Antibody, Biotin
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H302-Harmful if swallowed.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : SODIUM AZIDE
Your input is important to us. Please complete this form to provide feedback related to the content on this product.