missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgA/IgG/IgM (H+L) Secondary Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-73622
This item is not returnable.
View return policy
Description
IgA/IgG/IgM (H+L) Polyclonal specifically detects IgA/IgG/IgM (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgA/IgG/IgM (H+L) | |
| Polyclonal | |
| Biotin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| This reagent is suitable for the detection of all mouse immunoglobulin subclasses, isotypes and chain combinations. | |
| Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:2000 - 1:10000, ELISA 1:500000, Immunohistochemistry 1:1000 - 1:5000 | |
| Mouse IgG IgA and IgM whole molecule | |
| 2 mg | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction