missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgA Heavy Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 13 publications
Brand: Novus Biologicals NB7501
This item is not returnable.
View return policy
Description
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| IgA Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| Immunoglobulin A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3493 | |
| Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:30000, ELISA 1:1000-1:30000 (Primary) 1:100-1:500 (for coating plates), Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:200-1:2000, Immunohistochemistry-Paraffin 1:200-1:2000 | |
| IGHA1 | |
| Mouse IgA | |
| 1 mL | |
| Secondary | |
| By immunoelectrophoresis and ELISA this reacts specifically with mouse IgA. This may cross react with IgA from other species. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction