missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Monkey IgM Heavy Chain Secondary Antibody, Janelia Fluor 646, Novus Biologicals™

Product Code. 18423608 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423608 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423608 Supplier Novus Biologicals Supplier No. NBP259719JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM Heavy Chain
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain
Host Species Goat
Immunogen Monkey IgM whole molecule
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3507
Target Species Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.