missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Monkey IgG Heavy Chain Secondary Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-73557
This item is not returnable.
View return policy
Description
IgG Heavy Chain Polyclonal antibody specifically detects IgG Heavy Chain in Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG Heavy Chain | |
| Polyclonal | |
| Biotin | |
| Immunoglobulin G, ImmunoglobulinG | |
| Goat anti-Monkey IgG (gamma chain) Secondary Antibody was produced by repeated immunization with monkey IgG gamma heavy chain in goat. | |
| 1 mg | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:2000 - 1:5000, ELISA 1:20000 - 1:50000, Immunohistochemistry 1:1000 - 1:2500 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| No reaction was observed against Monkey IgA or Monkey IgM. | |
| Monkey | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction