missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Monkey IgA/IgG/IgM (H+L) Secondary Antibody, FITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69274
This item is not returnable.
View return policy
Description
IgA/IgG/IgM (H+L) Polyclonal antibody specifically detects IgA/IgG/IgM (H+L) in Monkey samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| IgA/IgG/IgM (H+L) | |
| Polyclonal | |
| FITC | |
| Lyophilized from PBS (pH 7.2) | |
| Purified polyclonal monkey IgG, and IgA and IgM containing factions isolated from monkey serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Secondary | |
| Monkey | |
| Lyophilized |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| LYOPH | |
| ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Goat | |
| Ion exchange chromatography | |
| RUO | |
| Reconstitute by by adding 1 mL sterile distilled water. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction