Learn More
MMP3 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MMP3.
Brand: Abnova PAB27568.100ug
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes an enzyme which degrades fibronectin, laminin, collagens III, IV, IX, and X, and cartilage proteoglycans. The enzyme is thought to be involved in wound repair, progression of atherosclerosis, and tumor initiation. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. [provided by RefSeq
Sequence: DPNAKKVTHTLKSNSSpecifications
| MMP3 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.1-0.3ug/ml) The optimal working dilution should be determined by the end user. | |
| MMP3 | |
| MMP3 | |
| A synthetic peptide corresponding to amino acids 459-473 at C-terminus of human MMP3. | |
| 100 μg | |
| Primary | |
| Human |
| Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of MMP3. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| CHDS6/MGC126102/MGC126103/MGC126104/MMP-3/SL-1/STMY/STMY1/STR1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4314 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only