Learn More
MGLL, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MGLL.
Brand: Abnova PAB7494.100ug
Description
Monoglyceride lipase (MGLL; EC 3.1.1.23) functions together with hormone-sensitive lipase (LIPE; MIM 151750) to hydrolyze intracellular triglyceride stores in adipocytes and other cells to fatty acids and glycerol. MGLL may also complement lipoprotein lipase (LPL; MIM 238600) in completing hydrolysis of monoglycerides resulting from degradation of lipoprotein triglycerides (Karlsson et al., 2001 [PubMed 11470505]).[supplied by OMIM
Sequence: C-QDLPHLVNADGQYSpecifications
| MGLL | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| MGLL | |
| MGLL | |
| A synthetic peptide corresponding to human MGLL. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_009214.1 and NP_0010.03794.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of MGLL. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| HU-K5/HUK5/MGL | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 11343 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only