Learn More
MDFI Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MDFI.
Brand: Abnova PAB17227.100ug
Description
This protein is a transcription factor that negatively regulates other myogenic family proteins. Studies of the mouse homolog, I-mf, show that it interferes with myogenic factor function by masking nuclear localization signals and preventing DNA binding. Knockout mouse studies show defects in the formation of vertebrae and ribs that also involve cartilage formation in these structures. [provided by RefSeq
Sequence: C-SKSTTSQIPLQAQEDSpecifications
| MDFI | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
| MDFI | |
| MDFI | |
| A synthetic peptide corresponding to amino acids at internal region of human MDFI. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of MDFI. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| I-MF/I-mfa | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 4188 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only