missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCH6. Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MARCH6.
Brand: Abnova PAB7609.100ug
This item is not returnable.
View return policy
Description
Sequence: C-QGRRVIFQKVKESpecifications
| MARCH6. | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.2-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| MARCH6 | |
| MARCH6 | |
| A synthetic peptide corresponding to amino acids 701-712 of human MARCH6. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of MARCH6. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| KIAA0597/MARCH-VI/RNF176/TEB4 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10299 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction