Learn More
MAP3K6, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MAP3K6.
Brand: Abnova PAB17236.100ug
Description
This gene encodes a member of the serine/threonine protein kinase family. The encoded kinase was identified by its interaction with MAP3K5/ASK, a protein kinase and an activator of c-Jun kinase (MAPK7/JNK) and MAPK14/p38 kinase. This kinase was found to weakly activate MAPK7, but not MAPK1/ERK or MAPK14. An alternatively spliced transcript variant has been found for this gene, but its biological validity has not been determined. [provided by RefSeq
Sequence: C-RAILAQRAGSTPSpecifications
| MAP3K6 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| MAP3K6 | |
| MAP3K6 | |
| A synthetic peptide corresponding to human MAP3K6. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of MAP3K6. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| ASK2/MAPKKK6/MGC125653/MGC20114 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 9064 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only