Learn More
MAGI2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MAGI2.
Brand: Abnova PAB6534.100ug
Description
The protein encoded by this gene interacts with atrophin-1. Atrophin-1 contains a polyglutamine repeat, expansion of which is responsible for dentatorubral and pallidoluysian atrophy. This encoded protein is characterized by two WW domains, a guanylate kinase-like domain, and multiple PDZ domains. It has structural similarity to the membrane-associated guanylate kinase homologue (MAGUK) family. [provided by RefSeq
Sequence: C-PSVLKPGASAASRSpecifications
| MAGI2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) The optimal working dilution should be determined by the end user. | |
| MAGI2 | |
| MAGI2 | |
| A synthetic peptide corresponding to human MAGI2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of MAGI2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| ACVRIP1/AIP1/ARIP1/MAGI-2/SSCAM | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 9863 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only