Learn More
KCNJ1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised aganist synthetic peptide of KCNJ1.
Brand: Abnova PAB15638.100ug
Description
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. It is activated by internal ATP and probably plays an important role in potassium homeostasis. The encoded protein has a greater tendency to allow potassium to flow into a cell rather than out of a cell. Mutations in this gene have been associated with antenatal Bartter syndrome, which is characterized by salt wasting, hypokalemic alkalosis, hypercalciuria, and low blood pressure. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: C-DQININFVVDAGNENSpecifications
| KCNJ1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| KCNJ1 | |
| KCNJ1 | |
| A synthetic peptide corresponding to amino acids at internal region of human KCNJ1. | |
| 100 μg | |
| Primary | |
| Approximately 45 KDa band observed in human, mouse and rat kidney lysates (calculated MW of 44.8 KDa according to NP_000211.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised aganist synthetic peptide of KCNJ1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| KIR1.1/ROMK/ROMK1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 3758 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only