Learn More
KCNC3 Goat anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of KCNC3.
Brand: Abnova PAB24567.100ug
Description
The Shaker gene family of Drosophila encodes components of voltage-gated potassium channels and is comprised of four subfamilies. Based on sequence similarity, this gene is similar to one of these subfamilies, namely the Shaw subfamily. The protein encoded by this gene belongs to the delayed rectifier class of channel proteins and is an integral membrane protein that mediates the voltage-dependent potassium ion permeability of excitable membranes. [provided by RefSeq
Sequence: C-HISNKTVTQASPSpecifications
| KCNC3 | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of KCNC3. | |
| In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| KSHIIID/KV3.3/SCA13 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 3748 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| KCNC3 | |
| KCNC3 | |
| A synthetic peptide corresponding to amino acids 317-328 at internal region of human KCNC3. | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only