Learn More
KALRN, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of KALRN.
Brand: Abnova PAB6507.100ug
Description
Huntington's disease (HD), a neurodegenerative disorder characterized by loss of striatal neurons, is caused by an expansion of a polyglutamine tract in the HD protein huntingtin. This gene encodes a protein that interacts with the huntingtin-associated protein 1, which is a huntingtin binding protein that may function in vesicle trafficking. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]
Sequence: C-RWHLGPGDPFSTYVSpecifications
| KALRN | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| KALRN | |
| KALRN | |
| A synthetic peptide corresponding to C-terminus of human KALRN. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize isoform 2 (NP_0.03938.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of KALRN. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| DUET/FLJ16443/HAPIP/TRAD/duo | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 8997 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only