Learn More
IQGAP1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of IQGAP1.
Brand: Abnova PAB18725.100ug
Description
This gene encodes a member of the IQGAP family. The protein contains four IQ domains, one calponin homology domain, one Ras-GAP domain and one WW domain. It interacts with components of the cytoskeleton, with cell adhesion molecules, and with several signaling molecules to regulate cell morphology and motility. Expression of the protein is upregulated by gene amplification in two gastric cancer cell lines. [provided by RefSeq
Sequence: C-KRQSGQTDPLQKEESpecifications
| IQGAP1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| IQGAP1 | |
| IQGAP1 | |
| A synthetic peptide corresponding to amino acids at internal region of human IQGAP1. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of IQGAP1. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| HUMORFA01/KIAA0051/SAR1/p195 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 8826 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only