Learn More
IDS, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of IDS.
Brand: Abnova PAB19674.100ug
Description
Iduronate-2-sulfatase is required for the lysosomal degradation of heparan sulfate and dermatan sulfate. Mutations in this X-chromosome gene that result in enzymatic deficiency lead to the sex-linked Mucopolysaccharidosis Type II, also known as Hunter Syndrome. Iduronate-2-sulfatase has a strong sequence homology with human arylsulfatases A, B, and C, and human glucosamine-6-sulfatase. A splice variant of this gene has been described. [provided by RefSeq
Sequence: C-KHFRFRDLEEDPSpecifications
| IDS | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.1-0.3 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| IDS | |
| IDS | |
| A synthetic peptide corresponding to internal region of human IDS. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize isoform a (NP_000193.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of IDS. | |
| In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| MPS2/SIDS | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 3423 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only