missing translation for 'onlineSavingsMsg'
Learn More

IDE Goat anti-Human, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of IDE.

Brand:  Abnova PAB7317.100ug

Product Code. 16080398

  • £432.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response. [provided by RefSeq

Sequence: C-QQYNFDRDNTE
Specifications

Specifications

IDE
Polyclonal
Unconjugated
ELISA (1:32000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user.
IDE
IDE
A synthetic peptide corresponding to human IDE.
100 μg
Primary
Human
Liquid
ELISA, Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of IDE.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
FLJ35968/INSULYSIN
Goat
Antigen affinity purification
RUO
3416
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only