missing translation for 'onlineSavingsMsg'
Learn More

Escherichia coli O157:H7, Goat, Polyclonal Antibody, Abnova™

Product Code. 16011007
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16011007 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16011007 Supplier Abnova Supplier No. PAB13942.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Goat polyclonal antibody raised against native Escherichia coli O157:H7.

Specifications

Antigen Escherichia coli O157:H7
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against native Escherichia coli O157:H7.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM phosphate buffer, 150mM NaCl, pH 7.4 (0.09% sodium azide)
Host Species Goat
Immunogen Native purified Escherichia coli of O and H antigens.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with E. coli O157:H7. Cross-reacts weakly with Salmonella sp. O antigens.
Target Species E. coli
Content And Storage Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. If aliquoted for long term storage fill volume should be equal to or greater than 50% of the nominal fill volume of the vial used.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.