missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human Kappa Light Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB7480
This item is not returnable.
View return policy
Description
Kappa Light Chain Polyclonal specifically detects Kappa Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Kappa Light Chain | |
| Polyclonal | |
| Unconjugated | |
| HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3514 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:30000, ELISA 1:1000-1:30000 (Primary) 1:100-1:500 (for coating plates), Immunohistochemistry 1:200- 1:2000, Immunocytochemistry/Immunofluorescence 1:200- 1:2000, Immunohistochemistry-Paraffin | |
| IGKC | |
| Human Kappa Free Light Chain | |
| 1 mL | |
| Secondary | |
| By immunoelectrophoresis and ELISA this reacts specifically with kappa free light chains. No was detected against lambda light chain or other serum proteins. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction