missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human Kappa Light Chain Secondary Antibody, DyLight 650 (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-75937
This item is not returnable.
View return policy
Description
Kappa Light Chain Polyclonal specifically detects Kappa Light Chain in Human samples. It is validated for Flow Cytometry, Immunomicroscopy.
Specifications
| Kappa Light Chain | |
| Polyclonal | |
| DyLight 650 | |
| HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
| Goat | |
| Affinity purified | |
| RUO | |
| 3514 | |
| Rehydrate with 0.55 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
| Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. |
| Flow Cytometry, Immunomicroscopy | |
| LYOPH | |
| Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000 | |
| IGKC | |
| Purified human Ig | |
| 0.5 mg | |
| Secondary | |
| Based on IEP, this antibody reacts with kappa and light chains on all human immunoglobulins. This antibody has been pre-adsorbed against mouse serum. | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction