missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Human Kappa Light Chain Secondary Antibody, DyLight 350, Novus Biologicals™

Product Code. 18415986 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415986 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415986 Supplier Novus Biologicals Supplier No. NBP175060UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Kappa Light Chain Polyclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kappa Light Chain
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot : 1:100-1:2000, ELISA : 1:100-1:2000, Immunohistochemistry : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500, Immunodiffusion, Immunoassay
Formulation 50mM Sodium Borate
Gene Alias HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809
Host Species Goat
Immunogen Purified Human Kappa light chain
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3514
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.