missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Human J Chain Secondary Antibody, Novus Biologicals™

Product Code. 18600429 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18600429 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18600429 Supplier Novus Biologicals Supplier No. NBP269766

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

J Chain Polyclonal antibody specifically detects J Chain in Human samples. It is validated for ELISA, Immunoprecipitation, Immunoelectrophoresis
TRUSTED_SUSTAINABILITY

Specifications

Antigen J Chain
Applications ELISA, Immunoprecipitation, Immunoelectrophoresis
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA, Immunoprecipitation 1:10 - 1:500, Immunoelectrophoresis
Formulation Whole antiserum
Gene Alias IgJ chain;Immunoglobulin J chain;immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides;J chain;Joining chain of multimeric IgA and IgM
Host Species Goat
Immunogen Human J chain is a polypeptide folded within the structure of the polymeric immunoglobulin, resulting in a very limited exposure of J chain antigenic determinants. The antiserum is raised against the isolated and purified J chain. J chains isolated from polymeric IgA and IgM are identical by criteria of composition, peptide maps and antigenicity; Human J chain is distinct from all other chain components of polymeric IgA and IgM. It has a unique primary structure as shown by sequence analyses. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Whole Antiserum
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Reconstitution Reconstitute the lyophilized antiserum by adding 0.5 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.