missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Human J Chain Secondary Antibody, FITC, Novus Biologicals™

Product Code. 18623637 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18623637 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18623637 Supplier Novus Biologicals Supplier No. NBP269761

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

J Chain Polyclonal antibody specifically detects J Chain in Human samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen J Chain
Applications ELISA, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate FITC
Dilution ELISA, Immunohistochemistry-Frozen, Immunocytochemistry
Formulation Lyophilized from PBS,(pH 7.2)
Gene Alias IgJ chain;Immunoglobulin J chain;immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides;J chain;Joining chain of multimeric IgA and IgM
Host Species Goat
Immunogen Human J chain is a polypeptide folded within the structure of the polymeric immunoglobulin, resulting in a very limited exposure of J chain antigenic determinants. The antiserum is raised against the isolated and purified J chain. J chains isolated from polymeric IgA and IgM are identical by criteria of composition, peptide maps and antigenicity; Human J chain is distinct from all other chain components of polymeric IgA and IgM. It has a unique primary structure as shown by sequence analyses. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Reconstitution Reconstitute by adding 1 mL sterile distilled water, spin down to remove insoluble particles.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.