missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgM F( c)5mu fragment, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB9298.1mg
This item is not returnable.
View return policy
Specifications
| Goat anti-Human IgM F( c)5mu fragment | |
| Polyclonal | |
| Goat anti-human IgG recognizes human IgM F(c)5mu fragment. This secondary antibody was purified using antigen affinity chromatography. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Human IgM F(c)5mu fragment | |
| 1 mg | |
| No reaction was observed against anti-Pepsin or anti-goat IgG F(c). Specificity was confirmed by ELISA at less than 1% cross reactivity against other human heavy or light chain isotypes. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities, pepsin digestion and chromatographic separation. | |
| Secondary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction