missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG (Heavy chain) Superclonal™ Secondary Antibody, HRP

Product Code. 17847382
Change view
Click to view available options
Quantity:
50 μL
250 μL
Unit Size:
250µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17847382 250 μL 250µL
17818192 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17847382 Supplier Invitrogen™ Supplier No. A56862

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Recombinant Superclonal Secondary Antibody

This Goat-anti-Human IgG (Heavy Chain), Superclonal Recombinant Secondary Antibody specifically detects human IgG Heavy chain with no cross-reactivity to light chain and other species immunoglobulins except moderate cross-reactivity towards Monkey IgG. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity, animal origin-free formulation, and lot-to-lot consistency. Superclonal™ Recombinant antibodies are a defined pool of recombinant monoclonal antibodies, manufactured by selecting and pooling individual clones to provide complementary, specific, and optimal epitope coverage to achieve enhanced performance. The sensitivity and specificity of each lot is confirmed using ELISA. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG (Heavy chain)
Applications ELISA, Western Blot
Classification Recombinant Superclonal
Concentration 1.0 mg/mL
Conjugate HRP
Formulation PBS with proprietary stabilizer and proprietary preservative; pH 7.4
Host Species Goat
Immunogen Purified Human IgG.
Purification Method Protein G
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.