missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG (H+L) Secondary Antibody, HRP

Product Code. 10746093
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10746093 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10746093 Supplier Invitrogen™ Supplier No. H10307

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Goat Polyclonal Secondary Antibody

Goat anti-Human IgG (H+L) Secondary Antibody for ELISA, Western Blot

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG (H+L)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate HRP
Description Goat anti-Human IgG (H+L) Secondary Antibody, 2mL, 0.5mg/mL Concentration, Clear, Liquid, HRP Conjugate, Goat Host Species, Human Species Reactivity, PBS with 4mg/mL BSA, 40% Glycerol Storage Buffer, 4°C Storage Requirements, For Flow Cytometry
Formulation PBS with 40% glycerol, 4mg/mL BSA and no preservative
Host Species Goat
Purification Method Purified
Quantity 2 mL
Regulatory Status ASR
Research Discipline Immunology
Chains or Fragments Whole antibody
Whole Molecule Yes
Primary or Secondary Secondary
Packaging 1 Each
Target Species Human
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Detection Method HRP, AP, fluorescence
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.