missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Human IgG (H+L) Secondary Antibody, DyLight 488 (Pre-adsorbed), Novus Biologicals™

Product Code. 18428815 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428815 0.5 mL 0.50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428815 Supplier Novus Biologicals Supplier No. NBP172729G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Western Blot, ELISA, Immunohistochemistry, Dot Blot
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Host Species Goat
Immunogen Human IgG whole molecule
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.