missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG (H+L) Secondary Antibody, DyLight 405, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7483V
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG (H+L) | |
| Polyclonal | |
| 50mM Sodium Borate | |
| This Goat anti-Human IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against human IgG. | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction