missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Human IgG Fc Secondary Antibody
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ H10500
This item is not returnable.
View return policy
Description
Goat anti-Human IgG Fc Secondary Antibody for Flow
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Human IgG Fc | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.1% sodium azide | |
| Purified | |
| ASR | |
| Whole antibody | |
| Secondary | |
| 0.1% Sodium azide | |
| 4°C, store in dark | |
| Liquid | |
| HRP, AP, fluorescence |
| Flow Cytometry | |
| 0.8mg/mL | |
| Goat anti-Human IgG Fc Secondary Antibody, 1mL, 0.8mg/mL Concentration, Clear, Liquid, Unconjugated Conjugate, Goat Host Species, Human Species Reactivity, PBS Storage Buffer, 4°C Storage Requirements, For Flow Cytometry | |
| Goat | |
| 1 mL | |
| Immunology | |
| Yes | |
| 1 Each | |
| Human | |
| Secondary Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction