Learn More
Goat anti-Human IgG Fc Recombinant Secondary Antibody, Invitrogen™
Goat Recombinant Monoclonal Secondary Antibody
£88.75 - £277.00
Specifications
Antigen | Human IgG Fc |
---|---|
Clone | 3H8L9 |
Concentration | 1.0 mg/mL |
Applications | ELISA, Immunocytochemistry, Peptide Array, Western Blot |
Classification | Recombinant Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17830761
|
Invitrogen
A5573550UL |
50 μL |
£88.75
50µL |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17820761
|
Invitrogen
A55735 |
250 μL |
£277.00
250µL |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Goat anti-Human IgG FC Recombinant Secondary Antibody specifically detects human IgG Fc with no cross-reactivity to human IgA or IgM or various species immunoglobulins. The sensitivity and specificity of each lot is confirmed using ELISA. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Human IgG Fc | |
1.0 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.4 | |
IgG | |
Protein G | |
Secondary Antibody |
3H8L9 | |
ELISA, Immunocytochemistry, Peptide Array, Western Blot | |
Unconjugated | |
Goat | |
Human | |
Purified Human IgG | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |